Learn More
Cole-Parmer™ PFA Compression Fittings
Click to view available options
For Use With:
0.5 in. NPT (F)
0.5 in. NPT (M), 0.75 in. NPT (F)
Unit Size:
Each
Specifications
Specifications
| Material | PFA |
| For Use With | 0.5 in. NPT (M), 0.75 in. NPT (F) |
| Packaging Quantity | 1 Pack |
| Product Type | Compression Fitting |