Learn More
Carbolite™ PF 200 Drying Ovens
Product Code. 12069912
Change view
Click to view available options
Dimensions (L x W x H):
470 x 665 x 470 mm
920 x 965 x 670 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12069912 470 x 665 x 470 mm Each
12049912 920 x 965 x 670 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12069912 Supplier Carbolite™ Supplier No. PF200230SN+TMPR38301
  • ONLINE EXCLUSIVE PRICE
    6055.00€
    4980.00€ / Pack of 1
    Save 1075.00€
    17% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
Certifications/Compliance BS EN 61010-2-010:2003
Capacity (Metric) 230 L
Temperature (Metric) 30°C to 300°C
Type Drying Oven
Display Digital
For Use With (Application) Fan convection provides greater temperature uniformity and faster recovery rates than natural convection ovens.
Material Stainless Steel
Max. Shelf Load 10/50 kg
Model PF60
No. of Shelves 15 (Accepted), 2 (Fitted)
Ports Cable entry
Safety Features Over-temperature protection
Temperature Stability ±0.2°C
Temperature Uniformity ±5°C
Dimensions (D x W x H) Interior 255 x 330 x 320 mm
Dimensions (L x W x H) 470 x 665 x 470 mm
Length (Metric) 470 mm
Width (Metric) 665 mm
Width (Metric) Interior 590 mm
Height (Metric) 470 mm
Height (Metric) Interior 750 mm
Depth (Metric) 670 mm
Depth (Metric) Interior 520 mm
Shipping Weight (Metric) 75 kg
Weight (Metric) 100 kg
Voltage 240 V
Wattage 1180 w