Learn More
Carbolite™ PF 200 Drying Ovens
Product Code. 12049912
Click to view available options
Dimensions (L x W x H):
470 x 665 x 470 mm
920 x 965 x 670 mm
Unit Size:
Each
Product Code. 12049912
Brand: Carbolite™ PF200230SN+&02R38
Only null left
Specifications
Certifications/Compliance BS EN 61010-2-010:2003
Capacity (Metric) 230 L
Temperature (Metric) 30°C to 300°C
Type Drying Oven
Display Digital
For Use With (Application) Fan convection provides greater temperature uniformity and faster recovery rates than natural convection ovens.
Insulation Low thermal mass insulation
Material Stainless Steel
Max. Shelf Load 10/50 kg
Model PF200
No. of Shelves 15 (Accepted), 2 (Fitted)
Ports Cable entry
Safety Features Over-temperature protection
Temperature Stability ±0.2°C
Temperature Uniformity ±5°C
Dimensions (D x W x H) Interior 750 X 590 X 520 mm
Dimensions (L x W x H) 920 x 965 x 670 mm
Length (Metric) 920 mm
Width (Metric) 965 mm
Width (Metric) Interior 590 mm
Height (Metric) 670 mm
Height (Metric) Interior 750 mm
Depth (Metric) 670 mm
Depth (Metric) Interior 520 mm
Shipping Weight (Metric) 75 kg
Voltage 240 V
Wattage 1180 w
Frequency 50/60 Hz