Learn More
Carbolite™ PF Fan Convection Laboratory Oven
Product Code. 12059169
Change view
Click to view available options
Capacity (Metric):
127 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12059169 127 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12059169 Supplier Carbolite™ Supplier No. PF120230SN+TMPR38301
  • ONLINE EXCLUSIVE PRICE
    5110.00€
    4200.00€ / Pack of 1
    Save 910.00€
    17% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Designed to use fan convection for improved temperature uniformity. Carbolite™ PF Fan Convection Laboratory Oven provides a faster recovery rate than natural convection.
  • Laboratory ovens of the Peak series 300°C are available as a PF fan convection model.
  • Equipped with the R38 digital PID temperature controller
  • Fan convection for rapid heating and recovery and excellent uniformity
  • Chemically resistant stainless steel liner
  • Two nickel-chrome plated wire shelves
  • Lever latch door and airtight silicone seal
  • Compliant with safety standard BS EN 61010-2-010:2003
  • Options (specify at time of order)
    • Over-temperature protection (recommended to protect valuable contents and for unattended operation)
    • A range of sophisticated digital controllers, multi-segment programmers and data loggers is available. These can be fitted with RS232, RS485 or Ethernet communications
    • Access port for independent thermocouple
    • Accessory shelves and runners
    • Heavy-duty reinforced base and shelves
    • Cable entry ports
    • Variable speed fan control
    • Stoving and curing options for extraction of small volumes of volatile solvents (not compatible with viewing window option)
    • Viewing window (not compatible with stoving and curing option)
    • Interior light (not compatible with stoving and curing option)
    • Air exhaust fan
    • Moisture extraction option (comprising sealed seams and air exhaust fan)
    • Stacking frame to enable units to be stacked one upon another
    • Lockable door
    • Door switch to isolate elements and fan
    • Fixed or castor mounted floor stands
    • Low voltage options for use below 220V
    • Routine spares kit
Specifications
Certifications/Compliance BS EN 61010-2-010:2003
Capacity (Metric) 127 L
Temperature (Metric) Ambient +30° to 300°C
Type Fan Convection Drying Oven
Display Digital
Material Stainless Steel
Max. Shelf Load 10 kg/shelf
Model PF120
No. of Shelves 2 fitted, 9 available
Ports Cable entry
Temperature Stability ±0.2°C
Temperature Uniformity ±5.0°C at 300°C
Dimensions (D x W x H) Interior 500 x 490 x 520 mm
Dimensions (L x W x H) 670 x 865 x 670 mm
Voltage 240 V