missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PEX10 (Human) Recombinant Protein
Human PEX10 full-length ORF ( AAH18198.1, 1 a.a. - 326 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005192-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAPAAASPPEVIRAAQKDEYYRGGLRSAAGGALHSLAGARKWLEWRKEVELLSDVAYFGLTTLAGYQTLGEEYVSIIQVDPSRIHVPSSLRRGVLVTLHAVLPYLLDKALLPLEQELQADPDSGRPLQGSLGPGGRGCSGARRWMRHHTATLTEQQRRALLRAVFVLRQGLACLQRLHVAWFYIHGVFYHLAKRLTGITYLRVRSLPGEDLRARVSYRLLGVISLLHLVLSMGLQLYGFRQRQRARKEWRLHRGLSHRRASLEERAVSRNPLCTLCLEERRHPTATPCGHLFCWECITAWCSSKAECPLCREKFPPQKLIYLRHYR
Specifications
AAH18198.1 | |
peroxisomal biogenesis factor 10 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PEX10 | |
GST |
5192 | |
Wheat germ expression system | |
10 μg | |
MGC1998, NALD, RNF69 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |