Learn More
IKA™ PETTE S Pipette Tips
Shop All IKA Products
Click to view available options
Volume (Metric):
200 μL
Unit Size:
Pack of 1920
Description
- 200 μL transparent pipette tips featuring calibration rings
- Designed for IKA™ PETTE Pipettes
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Color | Transparent |
| Features | Calibration Rings |
| Format | TipBox |
| Packaging | 20 Boxes (96 Tips per Box) |
| Quantity | 1920 Tips (Total) |
| Color Code | Yellow |
| Disposable | Single-use |
| For Use With (Equipment) | IKA PETTE Pipette |
| Product Line | PETTE S |