Learn More
IKA™ PETTE MacroKit
GREENER_CHOICE
Product Code. 17543606 Shop All IKA Products
Click to view available options
Volume (Metric):
100 to 10,000 μL
Unit Size:
Each
Product Code. 17543606
Brand: IKA™ 0010007641
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The IKA PETTE MacroKit pipette package covers a volume range of 100 to 10,000 μL, ideal for laboratories that are working in the milliliter volume range.
  • The package consists of three single channel pipettes with variable volumes in the macro-volume range, a complimentary stand that holds up to three pipettes and three pipette tip boxes. This makes it a convenient starter package for researchers.
  • Features:
    • Mechanical air-cushion pipettes manufactured in Germany
    • Fully automated calibration by robots
    • Volume-locking feature to prevent any accidental volume changes
    • Large and clearly visible display
    • Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
    • Durable and robust construction using high quality UV resistant and chemical resistant materials
    • Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
    • Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
TRUSTED_SUSTAINABILITY
Specifications
Description Air Displacement Piston-Operated Pipette
Autoclavable Autoclavable
Number of Channels 1
Product Line PETTE
Sterility Sterile
Type Pipette Package
Volume (Metric) 100 to 10,000 μL
For Use With (Application) Working In The Milliliter Volume Range
Includes Three Single Channel Pipettes With Variable Volumes In The Macro-Volume Range, One Complimentary Stand That Holds Up To Three Pipettes, And Three Pipette Tip Boxes