Learn More
IKA™ PETTE Classic Kit
Shop All IKA Products
Click to view available options
Volume (Metric):
2 to 1000 μL
Unit Size:
Each
Description
- The package consists of three single channel pipettes with variable volumes, a complimentary stand that holds up to three pipettes and two pipette tip boxes. This makes it a convenient starter package for researchers.
- Features:
- Mechanical air-cushion pipettes manufactured in Germany
- Fully automated calibration by robots
- Volume-locking feature to prevent any accidental volume changes
- Large and clearly visible display
- Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
- Durable and robust construction using high quality UV resistant and chemical resistant materials
- Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
- Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
Specifications
Specifications
| Description | Air Displacement Piston-Operated Pipette |
| Autoclavable | Autoclavable |
| Number of Channels | 1 |
| Product Line | PETTE |
| Sterility | Sterile |
| Type | Pipette Package |
| Volume (Metric) | 2 to 1000 μL |
| Includes | Three Single Channel Pipettes With Variable Volumes, One Complimentary Stand That Holds Up To Three Pipettes, And Two Pipette Tip Boxes |