Learn More
IKA™ PETTE Classic Kit
Product Code. 17533606 Shop All IKA Products
Click to view available options
Volume (Metric):
2 to 1000 μL
Unit Size:
Each
Product Code. 17533606
Brand: IKA™ 0010013011
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The IKA PETTE Classic Kit pipette package covers a volume range of 2 to 1000 μL, sufficient for fulfilling the needs of most laboratories.
  • The package consists of three single channel pipettes with variable volumes, a complimentary stand that holds up to three pipettes and two pipette tip boxes. This makes it a convenient starter package for researchers.
  • Features:
    • Mechanical air-cushion pipettes manufactured in Germany
    • Fully automated calibration by robots
    • Volume-locking feature to prevent any accidental volume changes
    • Large and clearly visible display
    • Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
    • Durable and robust construction using high quality UV resistant and chemical resistant materials
    • Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
    • Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
TRUSTED_SUSTAINABILITY
Specifications
Description Air Displacement Piston-Operated Pipette
Autoclavable Autoclavable
Number of Channels 1
Product Line PETTE
Sterility Sterile
Type Pipette Package
Volume (Metric) 2 to 1000 μL
Includes Three Single Channel Pipettes With Variable Volumes, One Complimentary Stand That Holds Up To Three Pipettes, And Two Pipette Tip Boxes