Learn More
ratiolab™ Petri Dishes
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 500
Description
- Uncoated
- With vents
Specifications
Specifications
| Color | Clear |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Packaging | Cardboard |
| Quantity | 25 Dishes/Pk., 20 Pk./Cs. |
| Dimensions (Dia. x H) | 90 x 15 mm |