Learn More
ratiolab™ Petri Dishes
Product Code. 15642267
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 500
Product Code. 15642267
Brand: ratiolab™ 9050100
Only null left
Add to basket
Add to basket
For the cultivation of cell cultures.
  • Uncoated
  • With vents
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Packaging Cardboard
Quantity 25 Dishes/Pk., 20 Pk./Cs.
Dimensions (Dia. x H) 90 x 15 mm