Learn More
- Home
- Products
- Facility Safety and Maintenance
- Electrical and Lighting Maintenance
- Brady™ PermaSleeve™ Yellow Heat Shrink Wire and Cable Labels for M6/M7 Printers
Brady™ PermaSleeve™ Yellow Heat Shrink Wire and Cable Labels for M6/M7 Printers
Product Code. 17849214
Change view
Click to view available options
Dimensions (LxW):
0.182 x 1 in.
0.182 x 1.75 in.
0.235 x 1 in.
0.235 x 1.75 in.
0.335 x 1 in.
0.335 x 1.75 in.
0.439 x 1 in.
0.439 x 1.75 in.
0.444 x 1 in.
0.444 x 1.75 in.
0.645 x 1 in.
0.645 x 1.75 in.
0.851 x 1 in.
0.851 x 1.75 in.
1.25 x 1 in.
1.25 x 1.75 in.
1.667 x 1 in.
1.667 x 1.75 in.
Wire Gauge:
12 to 4 AWG
16 to 8 AWG
2 to 500 MCM
20 to 10 AWG
22 to 16 AWG
28 to 20 AWG
6 to 0000 AWG
8 to 1 AWG
Diameter (English):
0.094 in.
0.125 in.
0.187 in.
0.25 in.
0.375 in.
0.5 in.
0.75 in.
1 in.
For Use With (Equipment):
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
M710, BMP71
Unit Size:
Pack of 100
Pack of 50
30 product options available for selection
| Product Code. | Dimensions (LxW) | Wire Gauge | Diameter (English) | For Use With (Equipment) | unitSize |
|---|---|---|---|---|---|
| 17849214 | 0.182 x 1 in. | 28 to 20 AWG | 0.094 in. | M710, BMP71 | Pack of 100 |
| 17841409 | 0.182 x 1 in. | 28 to 20 AWG | 0.094 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17888934 | 0.182 x 1.75 in. | 28 to 20 AWG | 0.094 in. | M710, BMP71 | Pack of 100 |
| 17851429 | 0.182 x 1.75 in. | 28 to 20 AWG | 0.094 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17869184 | 0.235 x 1 in. | 22 to 16 AWG | 0.125 in. | M710, BMP71 | Pack of 100 |
| 17821469 | 0.235 x 1 in. | 22 to 16 AWG | 0.125 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17821539 | 0.235 x 1.75 in. | 22 to 16 AWG | 0.125 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17829204 | 0.235 x 1.75 in. | 22 to 16 AWG | 0.125 in. | M710, BMP71 | Pack of 100 |
| 17849074 | 0.335 x 1 in. | 20 to 10 AWG | 0.187 in. | M710, BMP71 | Pack of 100 |
| 17811639 | 0.335 x 1 in. | 20 to 10 AWG | 0.187 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17808764 | 0.335 x 1.75 in. | 20 to 10 AWG | 0.187 in. | M710, BMP71 | Pack of 100 |
| 17861429 | 0.335 x 1.75 in. | 20 to 10 AWG | 0.187 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17818964 | 0.439 x 1 in. | 16 to 8 AWG | 0.25 in. | M710, BMP71 | Pack of 100 |
| 17898754 | 0.439 x 1.75 in. | 16 to 8 AWG | 0.25 in. | M710, BMP71 | Pack of 100 |
| 17861399 | 0.444 x 1 in. | 16 to 8 AWG | 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17861579 | 0.444 x 1.75 in. | 16 to 8 AWG | 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17831699 | 0.645 x 1 in. | 12 to 4 AWG | 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17809004 | 0.645 x 1 in. | 12 to 4 AWG | 0.375 in. | M710, BMP71 | Pack of 100 |
| 17851389 | 0.645 x 1.75 in. | 12 to 4 AWG | 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17868764 | 0.645 x 1.75 in. | 12 to 4 AWG | 0.375 in. | M710, BMP71 | Pack of 100 |
| 17861489 | 0.851 x 1 in. | 8 to 1 AWG | 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17839114 | 0.851 x 1 in. | 8 to 1 AWG | 0.5 in. | M710, BMP71 | Pack of 100 |
| 17881569 | 0.851 x 1.75 in. | 8 to 1 AWG | 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17899074 | 0.851 x 1.75 in. | 8 to 1 AWG | 0.5 in. | M710, BMP71 | Pack of 100 |
| 17801479 | 1.25 x 1 in. | 6 to 0000 AWG | 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17811399 | 1.25 x 1.75 in. | 6 to 0000 AWG | 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 100 |
| 17881509 | 1.667 x 1 in. | 2 to 500 MCM | 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 50 |
| 17889024 | 1.667 x 1 in. | 2 to 500 MCM | 1 in. | M710, BMP71 | Pack of 50 |
| 17851369 | 1.667 x 1.75 in. | 2 to 500 MCM | 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Pack of 50 |
| 17828834 | 1.667 x 1.75 in. | 2 to 500 MCM | 1 in. | M710, BMP71 | Pack of 50 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
Denna artikel kan inte returneras.
Se returpolicy
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- Industrial grade with high temperature, UV, chemical, fuel, and oil resistance
- Self-extinguishing labels
- Crisp, clear identification resistant to abrasions and fading
- 3:1 heat shrinkable labels fit snugly around wires for maximum insulation protection and permanent identification
- Made of B-342 polyolefin material
Specifikationer
Specifikationer
| Certifications/Compliance | ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) |
| Adhesive Type | Non-adhesive |
| Color | Yellow |
| Finish | Matte |
| Format | Die-cut |
| Material | Polyolefin |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Electrical Labeling, Wire and Cable Labeling |
| For Use With (Equipment) | M710, BMP71 |
| Quantity | 100 Labels |
| Shape | Rectangle |
| Temperature Range (English) | -67°F to +275°F |
| Temperature Range (Metric) | -55°C to +135°C |
| Wire Gauge | 28 to 20 AWG |
| Dimensions (LxW) | 0.182 x 1 in. |
| Length (English) | 0.182 in. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Height (Metric) | 4.62 mm |
| Diameter (English) | 0.094 in. |
| Diameter (Metric) | 2.39 mm |
| Alternate Part Number | M7-094-1-342-YL |
| Visa mer Visa mindre |
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?