Learn More
Brady™ PermaSleeve Wiremarker Sleeves for BMP51 and BMP53
Product Code. 11982752
Click to view available options
Color:
Black on White
Unit Size:
Each
Product Code. 11982752
Brand: Brady™ 143227
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
PermaSleeve Wiremarkers Sleeves provide a wire marking solution combining speed, ease of use, and permanence to create high quality labels.
PermaSleeve markers provide a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Self-extinguishing
  • Meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, an dMIL-STD-202 method 215K
  • UL224 recognized
Specifications
Color Black on White
For Use With (Equipment) BMP51 Label Printer, BMP53 Label Printer
Material Polyolefin
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 0.3 in.
Height (Metric) 8.5 mm
Format Cartridge, Ribbon
Certifications/Compliance 2011/65/EU and 2015/863
For Use With (Application) Electrical Labeling, Wire and Cable Labeling
Quantity 80