Learn More
Brady™ PermaSleeve Wiremarker Sleeves for BMP41, BMP51, and BMP53
Click to view available options
Color:
Black on White
Black on Yellow
Height (Metric):
6 mm
8.5 mm
11.2 mm
Width (Metric):
6 mm
8.5 mm
11.2 mm
Unit Size:
Each
Description
PermaSleeve markers provide a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
- Self-extinguishing
- Meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, an dMIL-STD-202 method 215K
- UL224 recognized
Specifications
Specifications
| Color | Black on White |
| For Use With (Equipment) | BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer |
| Material | Polyolefin |
| Length (English) | 0.08 in. |
| Width (English) | 0.24 in. |
| Length (Metric) | 2 mm |
| Width (Metric) | 6 mm |
| Height (English) | 0.24 in. |
| Height (Metric) | 6 mm |
| Format | Cartridge, Ribbon |
| Show More |