Learn More
Brady™ PermaSleeve™ Low Smoke Zero Halogen Heat-shrink Labels
Product Code. 17871789
Click to view available options
Dimensions (LxW):
11.15 mm (H) x 1.68 m (L)
16.38 mm (H) x 1.68 m (L)
5.97 mm (H) x 1.68 m (L)
8.51 mm (H) x 1.68 m (L)
Color:
White
Yellow
Unit Size:
Each
Product Code. 17871789
Brand: Brady™ 170952
Permanently mark wires and cables with the low-smoke, halogen-free B-7641 polyolefin label material. Pre-sized to fit most wire and cable assemblies. Print with a Brady™ handheld label maker.
  • Designed for mass-transit and marine applications where flame-retardant, low-smoke wire marking materials are required
  • Flame-retardant and resistant to temperatures from -22°F to 221°F (-30°C to 105°C)
  • Fast shrinking with a 2:1 shrink ratio; permanently adheres to wires quickly with a heat gun (Brady item PMS-HG-1)
Specifications
Certifications/Compliance BS-6853 (1999) Vehicle Category 1A; EN-45545-2, requirement 24, Vehicle Category HL3;Zero Halogen Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Non-adhesive
Color Yellow
Finish Matte
Format Continuous
Material Polyolefin
Product Type Label
Sterility Non-sterile
For Use With (Application) Wire and cable labelling
For Use With (Equipment) BMP41;BMP51;BMP53;M511;M410;M510
Quantity 1
Temperature Range (English) -67°F to 221°F
Temperature Range (Metric) -55°C to 105°C
Wire Gauge 16 AWG - 10 AWG
Dimensions (LxW) 8.51 mm (H) x 1.68 m (L)
Length (English) 5.5 ft.
Length (Metric) 1.68 m
Width (English) 0.335 in.
Width (Metric) 0.85 cm
Height (English) 0.335 in
Height (Metric) 8.51 mm
Alternate Part Number M4C-187-7641-YL