Learn More
Brady™ PermaSleeve™ Low Smoke Zero Halogen Heat-shrink Labels
Permanently mark wires and cables with the low-smoke, halogen-free B-7641 polyolefin label material. Pre-sized to fit most wire and cable assemblies. Print with a Brady™ handheld label maker.
Brand:  Brady™ 170954
Additional Details : Weight : 0.16100kg
 View more versions of this product
Product Code. 17811829
  • 117.74€ / Each
Estimated Shipment: 22-08-2024
Description
  • Designed for mass-transit and marine applications where flame-retardant, low-smoke wire marking materials are required
  • Flame-retardant and resistant to temperatures from -22°F to 221°F (-30°C to 105°C)
  • Fast shrinking with a 2:1 shrink ratio; permanently adheres to wires quickly with a heat gun (Brady item PMS-HG-1)
Specifications
Specifications
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Black on Yellow
Continuous
Labels
Wire and Cable Labeling
1 Tape/Cartridge
5.5 ft. x 0.439 in.
0.439 in.
Non-adhesive
Matte
Heat-shrink Polyolefin
Non-sterile
BMP41, BMP51, M511
-67°F to +221°F
5.5 ft.
1.1 cm