Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Product Code. 10708242
Click to view available options
Length (Metric):
10 m
20 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 10
Product Code. 10708242
Brand: Macherey-Nagel™ 723341.50
Only null left
Specifications
Diameter (Metric) 0.5 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Permabond™ FFAP
USP Type G25, G35
For Use With (Application) Free Carboxylic Acids Analysis, Free Carboxylic Acids
Temperature Range 220°C to 240°C
Film Thickness 0.25 μm
Length (Metric) 50 m
Packing Material Permabond™ FFAP
Quantity 1 Each