Learn More
Macherey-Nagel™ PERMABOND™ CW 20 M Capillary Columns, 0.32mm ID
Product Code. p-8015534
Click to view available options
Length (Metric):
10 m
25 m
30 m
50 m
60 m
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
Unit Size:
Each
Product Code. 10480122
Brand: Macherey-Nagel™ 723827.10
Only null left
Add to basket
Add to basket
Designed for use in gas chromatography. Macherey-Nagel™ PERMABOND™ CW 20 M Capillary Columns, 0.32mm ID are composed of 20,000 Da polyethylene glycol. These polar phase columns are recommended for solvent analysis and alcohols, and are suitable for use with aqueous solutions.
Recommended for solvent analysis and alcohols in gas chromatography, and suitable for use with aqueous solutions.
  • Columns feature 0.32mm inner diameter and 0.5mm outer diameter, with film options of 0.25μm, 0.35μm and 0.50μm thickness
  • Available in a variety of lengths: 10m, 25m, 30m, 50mm and 60m
  • Maximum temperature:
    • 220°C in long-term isothermal operations
    • 240°C in short isotherms in a temperature program
  • Similar phases: OPTIMA™ WAX, DB-Wax, Supelcowax, HP‑Wax, HP-INNOWAX, Rtx‑Wax, CP-Wax 52 CB, Stabilwax, 007-CW, BP20, AT-Wax, ZB-Wax
Specifications
Max. Temperature 220°C (long term), 240°C (in temperature program)
Diameter (Metric) 0.5 mm
Functionality/Interaction Type Polar phase
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Permabond™ CW 20 M
USP Type G16
For Use With (Application) Gas Chromatography (GC), Aqueous Solutions
Film Thickness 0.35 μm
Length (Metric) 10 m
Packing Material Poly(ethylene glycol)
Quantity 1 Each