Learn More
Perindopril L-Arginine, TRC
Product Code. 18083440
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083440 5 mg 5mg
18023850 1 mg 1mg
18081650 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18083440 Supplier Toronto Research Chemicals Supplier No. TRCP2876005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 612548-45-5
Molecular Formula C25H46N6O7
Molecular Weight (g/mol) 542.67
Synonym L-Arginine, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate (1:1), L-Arginine, mono[(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate] (9CI), (2S)-2-Amino-5-guanidinopentanoic acid (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]propanoyl]octahydro-1H-indole-2-carboxylate, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate L-Arginine (1:1), mono[(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate]
IUPAC Name (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;(2S)-2-amino-5-carbamimidamidopentanoic acid
SMILES CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)OCC.N[C@@H](CCCNC(=N)N)C(=O)O
Specifications
InChI Formula InChI=1S/C19H32N2O5.C6H14N4O2/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;7-4(5(11)12)2-1-3-10-6(8)9/h12-16,20H,4-11H2,1-3H3,(H,23,24);4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t12-,13-,14-,15-,16-;4-/m00/s1
Quantity 5 mg
Formula Weight 542.3428
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Perindopril L-Arginine
Grade HPLC