Learn More
STEMCELL Technologies Perhexiline Maleate
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Perhexiline Maleate is an inhibitor of the mitochondrial enzymes carnitine palmitoyltransferase 1 (CPT1; IC50 = 0.077 mM in rat heart; IC50 = 0.148 mM in rat liver) and CPT2 (IC50 = 0.079 mM in rat heart; Kennedy et al. 1996; Kennedy et al.) 2000)
- Perhexiline Maleate also modulates autophagy via mammalian target of rapamycin complex 1 (mTORC1) signaling (Balgi et al.)
- Alters myocardial metabolism from fatty acid to glucose utilization, resulting in higher ATP production and oxygen consumption (Ashrafian et al.; Jeffrey et al.)
- Inhibits mTORC1 signaling and induces autophagy in MCF-7 cells (Balgi et al.)
Specifications
Specifications
| CAS | 6724-53-4 |
| Quantity | 1 mg |
| Molecular Formula | C19H35N · C4H4O4 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.