Learn More
Perforated Basket for Steam Sterilizer Autoclaves
Product Code. 12663815
Encompass_Preferred
Change view
Click to view available options
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12663815 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12663815 Supplier Raypa Supplier No. CI75
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Depth (Metric) Outside 380 mm
For Use With (Equipment) AE-75 DRY and AE-110 DRY Sterilmatic steam sterilizer autoclaves
Height (Metric) Exterior 260 mm
Model CI-75
Weight (Metric) 2.5 kg
Material Stainless Steel