Learn More
Perfluorotetradecanoic Acid (50μg/mL in Methanol), TRC
Specifications
Specifications
| CAS | 376-06-7 |
| Molecular Formula | C14 H F27 O2 |
| Quantity | 1 mL |
| Synonym | Tetradecanoic acid, heptacosafluoro- (9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluorotetradecanoic acid, PFTeDA, Perfluoro-n-tetradecanoic acid, Perfluoromyristic acid, Perfluorotetradecanecarboxylic acid, Perfluorotetradecanoic acid |
| InChI Formula | InChI=1S/C14HF27O2/c15-2(16,1(42)43)3(17,18)4(19,20)5(21,22)6(23,24)7(25,26)8(27,28)9(29,30)10(31,32)11(33,34)12(35,36)13(37,38)14(39,40)41/h(H,42,43) |
| SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid |
| Molecular Weight (g/mol) | 714.11 |
| Formula Weight | 713.9545 |
| Concentration or Composition (by Analyte or Components) | Perfluorotetradecanoic acid 50 μg/mL |
| Show More |