Learn More
Perfluoroheptanesulfonic Acid Sodium Salt, TRC
Product Code. 30135422
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135422 50 mg 50mg
30136690 25 mg 25mg
30007646 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135422 Supplier Toronto Research Chemicals Supplier No. TRCP69819650MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21934-50-9
Molecular Formula C7 F15 O3 S . Na
Molecular Weight (g/mol) 472.1
Synonym Perfluoroheptane Sulfonic Acid Sodium, 1-Heptanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro-, sodium salt (8CI,9CI), Sodium perfluoroheptanesulfonate, 1-Heptanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro-, sodium salt (8CI,9CI), Sodium perfluoroheptanesulfonate, Perfluoroheptanesulfonate, sodium salt, PFHpS, sodium salt
IUPAC Name sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7HF15O3S.Na/c8-1(9,2(10,11)4(14,15)6(18,19)20)3(12,13)5(16,17)7(21,22)26(23,24)25;/h(H,23,24,25);/q;+1/p-1
Formula Weight 471.9226
Percent Purity >95%
Chemical Name or Material Perfluoroheptanesulfonic acid sodium salt
Grade HPLC