Learn More
Perfluorodecane Sulfonic Acid, TRC
Product Code. 30041329
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041329 25 mg 25mg
30041186 50 mg 50mg
30041305 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30041329 Supplier Toronto Research Chemicals Supplier No. TRCP28654025MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 335-77-3
Molecular Formula C10HF21O3S
Molecular Weight (g/mol) 600.14
Synonym 1-Decanesulfonic acid, heneicosafluoro- (6CI,8CI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heneicosafluoro-1-decanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,1 0-Heneicosafluorodecane-1-sulfonic acid, Perfluorodecanesulfonic acid, PFDS
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosafluorodecane-1-sulfonic acid
SMILES OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10HF21O3S/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)35(32,33)34/h(H,32,33,34)
Formula Weight 599.93
Percent Purity >95%
Chemical Name or Material Perfluorodecane sulfonic acid
Grade HPLC