Learn More
Perfluorobutanoic acid 13C4 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Perfluorobutanoic acid 13C4 |
| CAS | 1017281-29-6 |
| Solvent or Matrix | Methanol/Water |
| Molecular Formula | 13C4 H F7 O2 |
| Quantity | 1 mL |
| Synonym | 13C4-PFBA, Perfluorobutanoic Acid 13C4, Perfluorobutanoic acid 13C4, Perfluoro-n-[13C4 ]butanoic acid, MPFBA, Butanoic-1,2,3,4-13C4 acid, 2,2,3,3,4,4,4-heptafluoro- (ACI), 2,2,3,3,4,4,4-Heptafluorobutanoic-1,2,3,4-13C4 acid (ACI), Perfluoro-n-[13C4]butanoic acid |
| InChI Formula | InChI=1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)/i1+1,2+1,3+1,4+1 |
| SMILES | O[13C](=O)[13C](F)(F)[13C](F)(F)[13C](F)(F)F |
| IUPAC Name | 2,2,3,3,4,4,4-heptafluoro(1,2,3,4-^{13}C_{4})butanoic acid |
| Molecular Weight (g/mol) | 218.00898 |
| Show More |