Learn More
Perfluoro(2-methyl-3-pentanone), TRC
Product Code. 30409740
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409740 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30409740 Supplier Toronto Research Chemicals Supplier No. TRCP9992355G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Perfluoro-2-methyl-3-pentanone
CAS 756-13-8
Molecular Formula C6 F12 O
Quantity 5 g
Synonym 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Dodecafluoro-2-methylpentan-3-one, Heptafluoroisopropyl pentafluoroethyl ketone, Khaldon PFK 49, L 18543, Novec 1230, Novec 649, Perfluoro(ethyl isopropyl ketone), Perfluoro-(4-methylpentan-3-one), Perfluoro-2-methyl-3-pentanone
InChI Formula InChI=1S/C6F12O/c7-2(4(10,11)12,5(13,14)15)1(19)3(8,9)6(16,17)18
SMILES FC(F)(F)C(F)(F)C(=O)C(F)(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
Molecular Weight (g/mol) 316.04
Formula Weight 315.9758