Learn More
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%
Product Code. 11399285
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11399285
Brand: Thermo Scientific Alfa Aesar L16614.14
Request Bulk or Custom Format
CAS: 27639-98-1 | C12F24O4 | 664.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27639-98-1
Molecular Formula C12F24O4
Molecular Weight (g/mol) 664.09
MDL Number MFCD00084743
InChI Key IIYQDKPSLAINNX-UHFFFAOYSA-N
Synonym perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propoxy, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, propanoyl fluoride,2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, acmc-20ajy8, 3,6,9-trioxadodecanoyl fluoride, tetradecafluoro-2,5,8-tris trifluoromethyl
PubChem CID 119616
IUPAC Name 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propanoyl fluoride
SMILES C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)F
Specifications
Density 1.8
Boiling Point 158°C to 161°C
Flash Point None
Refractive Index 1.3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 664.09
Percent Purity 97%
Chemical Name or Material Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride