Learn More
Honeywell™ PERFECT POLY™ Skin Gloves
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
Description
- Polyamide knitted glove
- Gray polyurethane coating on palm and tips of fingers
- Elastic wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand as well as outstanding dexterity
- The glove offers maximum comfort and added sturdiness
- The ultra tight weave gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and ultra light coating optimise the glove's breathability
- Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
- Non pilling, the polyamide is suitable for precision work
- The gloves are certified as silicone-free to ensure their use in painting
- Blue color and gray coating make the glove more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standads, EN 388-2016 Mechanical hazards |
| Description | 18 gauge |
| Color | Blue, Gray |
| Material | Polyamide, Polyurethane |
| Size | 7 |
| Length (Metric) | 24 cm |