Learn More
Hecht Karl™ Percisely Adjusted Specific Gravity Bottle
Product Code. 10043821
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10043821 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10043821 Supplier Hecht Karl™ Supplier No. 42571024
  • ONLINE EXCLUSIVE PRICE
    79.00€
    53.52€ / Pack of 1
    Save 25.48€
    32% Off
View all promotions
Estimated Shipment: 27-11-2026
Specific grafity bottles (pycnometers) acc. Gay-Lussac, with ground-in stopper.
Specifications
Quantity 1Each
Type Specific Gravity Bottle
Capacity (Metric) 25 mL
Product Type Specific Gravity Bottle