Learn More
Pentanedioic-d6 Anhydride, CDN
Product Code. 30312368
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312368
Brand: CDN CDND61620.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pentanedioic-D6 Anhydride
CAS 1219794-53-2
Molecular Formula C5 D6 O3
Quantity 0.5 g
Synonym Dihydro-2 H-pyran-2,6(3 H)-dione-3,3,4,4,5,5-d6, Glutaric anhydride-d6, Glutaric-d6 acid anhydride, Oxacyclohexane-2,6-dione-d6, Pentanedioic-2,2,3,3,4,4-d6 acid anhydride, Glutaric-d6 anhydride
InChI Formula InChI=1 S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2/i1D2,2D2,3D2
SMILES [2 H]C1([2 H])C(=O)OC(=O)C([2 H])([2 H])C1([2 H])[2 H]
IUPAC Name 3,3,4,4,5,5-hexadeuteriooxane-2,6-dione
Molecular Weight (g/mol) 120.1363
Formula Weight 120.0694 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity