missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylenebis(triphenylphosphonium bromide), 98+%, Thermo Scientific™

Product Code. p-7043494
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11324414

Brand: Thermo Scientific Alfa Aesar L00997.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pentamethylenebis(triphenylphosphonium bromide), is used as an intermediate in Pharmaceutical research.

Solubility
It is soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. Moisture sensitive. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-31-7
Molecular Formula C41H40Br2P2
Molecular Weight (g/mol) 754.53
MDL Number MFCD00051887
InChI Key XIZMFZWZYVQIOX-UHFFFAOYSA-L
Synonym pentane-1,5-diylbis triphenylphosphonium bromide, pentamethylenebis triphenylphosphonium bromide, triphenyl 5-triphenylphosphaniumylpentyl phosphanium dibromide, triphenyl 5-triphenylphosphaniumyl pentyl phosphanium dibromide, 1,5-pentanediylbis triphenylphosphonium dibromide, 1,1'-1,5-pentanediyl bis 1,1,1-triphenyl-, bromide 1:2
PubChem CID 3084382
IUPAC Name triphenyl(5-triphenylphosphaniumylpentyl)phosphanium;dibromide
SMILES [Br-].[Br-].C(CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 253°C to 260°C
Quantity 5 g
Beilstein 3875042
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 754.54
Percent Purity ≥98%
Chemical Name or Material Pentamethylenebis(triphenylphosphonium bromide)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.