Learn More
Pentafluorophenyl isothiocyanate, 96%
Product Code. 11322687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322687 1 g 1g
11332687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11322687 Supplier Thermo Scientific Alfa Aesar Supplier No. L11874.03
Request Bulk or Custom Format
CAS: 35923-79-6 | C7F5NS | 225.136 g/mol
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35923-79-6
Molecular Formula C7F5NS
Molecular Weight (g/mol) 225.136
MDL Number MFCD00041042
InChI Key NGNKMRBGZPDABE-UHFFFAOYSA-N
Synonym pentafluorophenyl isothiocyanate, perfluorophenyl isothiocyanate, pentafluorophenylisothiocyanate, benzene, pentafluoroisothiocyanato, 2,3,4,5,6-pentafluorophenylisothiocyanate, 2,3,4,5,6-pentafluorophenyl isothiocyanate, acmc-20ajyt, pubchem5009, pentafluorophenyl isothiocyanate 1g, 2,3,4,5,6-pentafluorobenzenisothiocyanate
PubChem CID 123452
IUPAC Name 1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
SMILES C(=NC1=C(C(=C(C(=C1F)F)F)F)F)=S
Specifications
Density 1.56
Boiling Point 109°C to 110°C (35 mmHg)
Refractive Index 1.5378
Quantity 1 g
UN Number UN2922
Beilstein 2736207
Sensitivity Moisture sensitive
Formula Weight 225.14
Percent Purity 96%
Chemical Name or Material Pentafluorophenyl isothiocyanate