Learn More
Pentachlorophenyl Laurate, TRC
Specifications
Specifications
| Chemical Name or Material | Pentachlorophenol dodecanoate |
| CAS | 3772-94-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H23 Cl5 O2 |
| Quantity | 10 mg |
| Synonym | Dodecanoic acid','2,3,4,5,6-pentachlorophenyl ester, Dodecanoic acid','pentachlorophenyl ester (9CI), Lauric acid','pentachlorophenyl ester (6CI,7CI,8CI), Phenol','pentachloro-','laurate (8CI), Lauroyl pentachlorophenate, Mystox, Mystox LPL, Mystox LSD, Mystox LSL, NSC 77542, Pentachlorophenol laurate, Pentachlorophenyl laurate |
| InChI Formula | InChI=1S/C18H23Cl5O2/c1-2-3-4-5-6-7-8-9-10-11-12(24)25-18-16(22)14(20)13(19)15(21)17(18)23/h2-11H2,1H3 |
| SMILES | CCCCCCCCCCCC(=O)Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl |
| IUPAC Name | (2,3,4,5,6-pentachlorophenyl) dodecanoate |
| Molecular Weight (g/mol) | 448.64 |
| Show More |