Learn More
Pentachloronitrobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | Quintozene |
| CAS | 82-68-8 |
| Molecular Formula | C6 Cl5 N O2 |
| Quantity | 25 g |
| Synonym | Quintozene, Pentachloronitrobenzene, Benzene','1,2,3,4,5-pentachloro-6-nitro-, Benzene','pentachloronitro- (8CI,9CI), 1,2,3,4,5-Pentachloro-6-nitrobenzene, 1,2,3,4,5-Pentachloronitrobenzene, 2,3,4,5,6-Pentachloronitrobenzene, Avicol (pesticide), Batrilex, Botrilex, Brassicol, Brassicol 75, Brassicol Super, Chinozan, Fartox, Folosan, Fomac 2, GC 3944-3-4, Kobutol, Korsikol, Marisan forte, NSC 58427, Nitropentachlorobenzene, PCNB, Pentagen, Phomasan, Plantacol, Quinosan, Quintozen, RTU 1010, RTU PCNB, Terrachlor, Terraclor, Terraclor 30G, Terrafun, Terrazan, Terrazan F, Tilcarex, Tritisan |
| InChI Formula | InChI=1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9 |
| SMILES | [O-][N+](=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl |
| IUPAC Name | 1,2,3,4,5-pentachloro-6-nitrobenzene |
| Molecular Weight (g/mol) | 295.33 |
| Formula Weight | 292.8372 |