missing translation for 'onlineSavingsMsg'
Learn More

Melford Penicillin V Potassium Salt

Product Code. 30018689
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018689 25 g 25g
30018688 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018689 Supplier Melford Supplier No. P3300025.0
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibitor of bacterial cell wall synthesis.

Inhibitor of bacterial cell wall synthesis. Active against gram positive and some gram negative bacteria. B-Lactamase sensitive.

Specifications

Chemical Name or Material Penicillin V Potassium Salt
CAS 132-98-9
Quantity 25 g
Molecular Formula C16H17KN2O4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.