Description
- Stainless steel AISI 304 (1.4301)
- For industry and water treatment
Specifications
Specifications
| Product Type | Dipper Beaker |
| Material | Stainless Steel (AISI 304 (1.4301)) |
| Autoclavable | Autoclavable |
| Capacity (English) | 33.81 oz. |
| Capacity (Metric) | 1000 mL |
| For Use With (Application) | Deep and Narrow Shafts |
| For Use With (Equipment) | Telescopic Rod |
| Height (English) | 6.42 in. |
| Height (Metric) | 163 mm |
| Length (English) | 7.91 in. |
| Show More |