Learn More
Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™
Product Code. p-7236577 Shop All MP Biomedical Products
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
Product Code. 17368573
Brand: MP Biomedicals 091670246
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pen-Strep (10,000 IU/mL, 10 mg/mL) is a double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
  • This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Concentration 10,000 IU/mL Penicillin, 10 mg/mL Streptomycin
Quantity 20 mL
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.