Learn More
3M™ PELTOR™ ProTac™ III Headset
Product Code. p-7083201
Click to view available options
Model:
PELTOR™ ProTac III Headset
PELTOR™ ProTac III Slim Headset
Style:
Hard Hat
Headband
Noise Reduction Rating (NRR):
19 dB, 21 dB
23 dB
26 dB
Unit Size:
Each
Product Code. 15638175
Brand: 3M™ 7100088424
Only null left
Add to basket
Edge
Add to basket
Headset with level-dependent function for ambient listening helps you focus on your work and communicate with colleagues in noisy environments, without removing the hearing protection. Stereo feedback allows one to better determine direction of alarms, warning cries, backing vehicles and so on.
  • Hearing protector helps protect against harmful noise, whilst allowing ambient sounds to be heard
  • Built-in level-dependent function allows the wearer to hear ambient sounds, such as conversations, machine sounds and warning signals
  • 3.5 mm stereo jack to connect to your mobile phone or two-way radio to hear the call right in your ear
  • Auto power off turns headset off after 4 hours of non-use, to save energy and a low-battery warning informs you when to change batteries
  • Voice guided menu system so no need to take your headset off
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Hearing Protection Style Earmuffs
For Use With (Application) Agriculture
Model PELTOR™ ProTac III Headset
Noise Reduction Rating (NRR) 26 dB
Product Type Protective Hearing Communications Headset
Style Headband