Learn More
3M™ PELTOR™ LiteCom™ Plus Headset
Product Code. p-7160319
Click to view available options
Unit Size:
Each
Model:
Helmet Mounted
Neckband
Product Code. 15779997
Brand: 3M™ 7000108543
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
3M™ PELTOR™ LiteCom™ Plus Headset is based on a robust basic design with a number of added features.
  • Headset with built in communication radio allows two-way conversation up to 3000 meters
  • Level dependent function lets the user hear surrounding noise and converse with colleagues
  • It's easy to connect a mobile phone or external communication radio, regardless of the frequency
  • Microphone software is programmed only to pick up speech and effectively filters out all other sounds
  • 121 sub-channels allow several users to communicate individually using the same channel
  • Communication on 8 channels in the PMR 446MHz band
  • Up to 1 kilometer range
TRUSTED_SUSTAINABILITY
Specifications
Product Type Communication Headset
Noise Level 33 dB
Certifications/Compliance CE Approved