Learn More
3M™ Peltor™ G300 Forestry Hard Hat Combination
Product Code. 17931030
Click to view available options
:
Color: Orange
Product Code. 17931030
Brand: 3M™ 7000104017
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The 3M™ Peltor™ G3000 Forestry Helmet Combination offers effective protection for the head, face and hearing.
  • G3000 Hard Hat
  • Mesh or polycarboate visor or faceshield
  • Effective protection for the head, face and hearing
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance None
Color Orange
For Use With (Equipment) Earmuffs, Faceshield
Product Type Headgear Combination
Size Standard
Ventilated Not Vented
Head Style Front Brim