Learn More
Memmert™ Refrigerated Incubator with Peltier Cooling, 1060 L, Stainless Steel
Shop All Memmert ProductsDescription
- Intuitive and easy-to-use operating menu
- Almost exclusive use of high-quality, corrosion-resistant and easily cleanable stainless steel for the working chamber and housing
- Heating concept specifically adapted for a precise and homogenous temperature control
- A wide range of options for programming and documentation using interfaces, integrated data logger and Software AtmoCONTROL
- Double doors standard for all models: Prevention of contamination and drops in temperature, and at the same time an optimum view of the sensitive load through wide-area interior glass doors
- Fans integrated in the Peltier elements ensure rapid energy transfer and optimum temperature distribution
Specifications
Specifications
| Capacity (Metric) | 1060 L |
| Chamber Material | Stainless Steel |
| Material | Stainless Steel, Zinc-plated Steel |
| Temperature Range (Metric) | 0°C to 70°C |
| Type | Refrigerated Incubator |
| Door Style | Inner glass doors |
| Includes | 2 stainless steel electropolished grids |
| Model | IPP1060 |
| Temperature Accuracy | 0.1°C |
| Dimensions (D x W x H) Interior | 850 x 1040 x 1200 mm |
| Show More |