Learn More
Macherey-Nagel™ Pehanon™ pH Paper
Product Code. 10708911 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
1 to 12
1 to 2.8
10.5 to 13
2.8 to 4.6
3.8 to 5.5
4 to 9
5.2 to 6.8
6 to 8.1
7.2 to 8.8
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10708911 6 to 8.1 Pack of 200
11366464 3.8 to 5.5 Pack of 200
10004760 10.5 to 13 Pack of 200
10196160 5.2 to 6.8 Pack of 200
10547502 4 to 9 Pack of 200
10058100 2.8 to 4.6 Pack of 200
10736991 8 to 9.7 Pack of 200
11817043 1 to 12 Pack of 200
10460431 1 to 2.8 Pack of 200
10741601 7.2 to 8.8 Pack of 200
10687901 9.5 to 12 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10708911 Supplier Macherey-Nagel™ Supplier No. 90417
  • ONLINE EXCLUSIVE PRICE
    18.65€
    16.33€ / Pack of 200
    Save 2.32€
    12% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Measure pH in colored solutions. Pehanon™ pH paper puts the indicator and reference on one strip making it perfect for pH determinations in colored solutions.
PEHANON™ is a special pH-test strips that unifies the pH-indicator and the reference colors on one strip and has the following advantages:
Any sample color has the same effect on both, the reference colors and the reactive pad. This ensures unadulterated readings in colored solutions. The user can be sure to get accurate pH-values.
An invisible hydrophobic barrier just above the top color field prevents capillary action of the test solution beyond. The handle will remain dry and clean whatever the sample is. Therefore the use of the strip is very safe.
pH-values can be determined without a separate color chart. Workers and machinists can use single strips instead of complete packs with color chart which makes PEHANON™ highly economic.
Specifications
pH Range 6 to 8.1
Format Stick
For Use With (Application) Determination of pH in the colored solutions
Graduations 6.0, 6.3, 6.6, 6.9, 7.2, 7.5, 7.8, 8.1
Quantity 200 Pack
Type pH Paper