missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Peel and Present Dispensing Edge
Click to view available options
Color:
Black
Unit Size:
Each
Description
- Peel and Present.
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | BBP11 Label Printer, BBP12 Label Printer |
| Description | Peel and Present |
| Quantity | 1 Ea. |
| Alternate Part Number | BBP12/BBP11-PLnPR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction