Learn More
Cole-Parmer™ PEEK Chromatography HPLC Valve
Product Code. 15382994 Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric) Outer:
3.17 mm
Unit Size:
Each
Product Code. 15382994
Brand: Cole-Parmer™ 0201401
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material PEEK
Max. Pressure 34.4 bar
Type Chromatography HPLC Valve
Model P-733
Diameter (Metric) Outer 3.17 mm