Learn More
Heinz Herenz Pediatric Tip Flocked VIR-Swab Virus Flocked Swab
Description
Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.
- The liquid medium allows the division of the sample material and thus several tests from one sample.
- The ready to use devices are supplied in a peelpack which is easy to open.
- The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
- The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
- There are labels on the test tubes for a accurate sample identification.
- The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
- Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.
Specifications
Specifications
| Product Type | Universal Viral Transport |
| Format | Swab |
| Media Type | Neutralizers |
| Sterility | Sterile |
| Color | White |