missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pectinase, Aspergillus niger

Product Code. 15445659
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15445659

Brand: Thermo Scientific Chemicals J63408.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.

Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.