Learn More
Lenz Laborglasinstrumente™ Pear-shaped Evaporating Flask with Conical Ground Joint
Product Code. 11836144
Change view
Click to view available options
Volume (Metric):
10 mL
25 mL
50 mL
3000 mL
Neck Finish:
14/23
29/32
39/32
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Neck Finish Unit Size
11836144 3000 mL 29/32 Each
11973381 10 mL 14/23 Pack of 10
11963381 25 mL 14/23 Pack of 10
11983381 50 mL 14/23 Pack of 10
11876084 50 mL 39/32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11836144 Supplier Lenz Laborglasinstrumente™ Supplier No. 03042976
  • ONLINE EXCLUSIVE PRICE
    150.00€
    106.96€ / Pack of 1
    Save 43.04€
    28% Off
Estimated Shipment: 15-12-2026
Especially for rotary evaporators, with center neck and conical joint, pear-shaped, with inscription label.
Specifications
Material Duran Glass
Type Evaporating Flask
For Use With (Equipment) Rotary Evaporators
Neck Finish 29/32
Shape Pear
Volume (Metric) 3000 mL
Diameter (Metric) 175 mm