Specifications
| Antigen | Peanut Agglutinin |
| Concentration | 1 mg/mL |
| Quantity | 1 mg |
| Content And Storage | Store at 4°C in the dark. |
| Product Line | Novus |
| For Use With (Application) | A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use. The recommended concentration range for use is 5-20 μg/mL. |
| Buffer | 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM CaCl2, proprietary stabilizer |