Learn More
Chemsplash PE Machine Made Disposable Over Sleeve
GREENER_CHOICE
Product Code. p-200105879
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 2000
Product Code. 30058310
Brand: Chemsplash 2527.300.999
Only null left
Add to basket
Add to basket
Disposable Polyethylene Oversleeves are made from 15 micron polyethylene material elasticated at both ends for ease of fit.
Disposable Polyethylene Oversleeves
  • 15 Micron Material
  • Elasticated at Both Ends
  • 40 x 20 cm
  • Heat Sealed Seams
  • PFAS Free
Description: Disposable Polyethylene Oversleeves are made from 15-micron polyethylene material, elasticated at both ends for ease of fit.
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Polyethylene
Color Blue
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Welded
Latex Free Yes
Length (English) 16 in.
Length (Metric) 16 cm