missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PDE6D (Human) Recombinant Protein
Human PDE6D full-length ORF ( AAH07831, 1 a.a. - 150 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005147-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSAKDERAREILRGFKLNWMNLRDAETGKILWQGTEDLSVPGVEHEARVPKKILKCKAVSRELNFSSTEQMEKFRLEQKVYFKGQCLEEWFFEFGFVIPNSTNTWQSLIEAAPESQMMPASVLTGNVIIETKFFDDDLLVSTSRVRLFYV
Specifications
AAH07831 | |
phosphodiesterase 6D, cGMP-specific, rod, delta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDE6D | |
GST |
5147 | |
Wheat germ expression system | |
25 μg | |
PDED | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |