missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PDD 00017272

Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
PDD 00017272 is a potent poly (ADP ribose) glycohydrolase (PARG) inhibitor (IC50 values are 4.8 nM in biochemical assay and 9.2 nM in cell based assay).
Specifications
Specifications
| Chemical Name or Material | 1-[(2,4-Dimethyl-5-thiazolyl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide |
| CAS | 1945950-20-8 |
| Quantity | 1 mg |
| Target | PARG Inhibitors |
| Molecular Formula | C23H25N5O4S3 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction